Orders LinkContactUsLinkComponent
Labels for BradyJet J2000 Lab printer
Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-179-7425J, PP, White, J2000-Lab, 25×25 mm, 1500 labels/roll
Catalog # BRDY150025
Supplier:   Brady
Labels for BradyJet J2000 Lab printer
Catalog # BRDY150025
Supplier:  Brady
Labels for BradyJet J2000 Lab printer
Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-179-7425J, PP, White, J2000-Lab, 25×25 mm, 1500 labels/roll
Price
162,02 €
Log in for availability
1 * 1 Roll
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
  • Designed to stay attached and remain legible
  • Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Specifications
  • Printer model:
    J2000-Lab
  • Type:
    B-7425J
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    25×25 mm
  • Material:
    PP
  • Seq:
    07
  • Model:
    J20-179-7425J
  • Packed:
    1500 labels/roll