Accessories for laboratory hoods, Protector®
SpillStopper™ work surface, right rear cupsink cutout, 1829×762×32 mm, 93 kg
Catalog # LBCP9500601
Supplier: LABCONCO
Jump to:
Product Details & Documents
Specifications
- Description:SpillStopper™ work surface, right rear cupsink cutout, 1829×762×32 mm, 93 kg
- Seq:19