Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
ANALYTICAL ACHIRAL COLUMN DAICEL DCpak®PBT 150 x 4,6 mm 3 µm SFC 1 * 1 items
DAICAAS24
:
CHIRAL TECHNOLOGIES
ANALYTICAL ACHIRAL COLUMN DAICEL DCpak®PBT 150 x 4,6 mm 3 µm SFC 1 * 1 items
DAICAAS24
:
CHIRAL TECHNOLOGIES