Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Microslide superfrost cut pink glass cello ISO 8037/1 1,0mm 1 * 50 items
Catalog # MENZAA00008232E011
Supplier:
Epredia
Expand image
Microslide superfrost cut pink glass cello ISO 8037/1 1,0mm 1 * 50 items
Catalog # MENZAA00008232E011
Supplier:
Epredia