Orders LinkContactUsLinkComponent
VWR® Splash Heads, Stutzer, Straight
VWR® Splash Heads, Stutzer, Straight
29/32 NS, 14/23 NS
Catalog # 201-1399
Supplier:   VWR Collection
VWR® Splash Heads, Stutzer, Straight
Catalog # 201-1399
Supplier:  VWR Collection
VWR® Splash Heads, Stutzer, Straight
VWR® Splash Heads, Stutzer, Straight
29/32 NS, 14/23 NS
Price
68,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    29/32 NS
  • Seq:
    02