ContactUsLinkComponent
A EPAC2 ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Double Antibod y 1 * 1 KIT
Catalog # GENOIT8194
Supplier:  G-Biosciences
A EPAC2 ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Double Antibod y 1 * 1 KIT
Catalog # GENOIT8194
Supplier:  G-Biosciences