ContactUsLinkComponent
A COLI (Collagen Type I) ELISA with a senstivity of 0.188ng/ml and a range of 0.313-20ng/ml. Uses the detection method Sandwich ELIS A, Double Antibody 1 * 1 KIT
Catalog # GENOIT8190
Supplier:  G-Biosciences
A COLI (Collagen Type I) ELISA with a senstivity of 0.188ng/ml and a range of 0.313-20ng/ml. Uses the detection method Sandwich ELIS A, Double Antibody 1 * 1 KIT
Catalog # GENOIT8190
Supplier:  G-Biosciences