ContactUsLinkComponent
A P311 (Neuronal protein 3.1) ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8173
Supplier:  G-Biosciences
A P311 (Neuronal protein 3.1) ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8173
Supplier:  G-Biosciences