ContactUsLinkComponent
A DSP (Dentin Sialoprotein) ELISA with a senstivity of 0.469ng/ml and a range of 0.781-50ng/ml. Uses the detection method Sandwich E LISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8161
Supplier:  G-Biosciences
A DSP (Dentin Sialoprotein) ELISA with a senstivity of 0.469ng/ml and a range of 0.781-50ng/ml. Uses the detection method Sandwich E LISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8161
Supplier:  G-Biosciences