ContactUsLinkComponent
A IL-31 (interleukin-31) ELISA with a senstivity of 4.688pg/ml and a range of 7.813-500pg/ml. Uses the detection method Sandwich ELI SA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8277
Supplier:  G-Biosciences
A IL-31 (interleukin-31) ELISA with a senstivity of 4.688pg/ml and a range of 7.813-500pg/ml. Uses the detection method Sandwich ELI SA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8277
Supplier:  G-Biosciences