ContactUsLinkComponent
An ELISA kit for the detection of TAT (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375pg/ml. 1 * 1 KIT
Catalog # GENOIT6009
Supplier:  G-Biosciences
An ELISA kit for the detection of TAT (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375pg/ml. 1 * 1 KIT
Catalog # GENOIT6009
Supplier:  G-Biosciences