ContactUsLinkComponent
An ELISA kit for the detection of TAFI (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.469ng/ml. 1 * 1 KIT
Catalog # GENOIT6007
Supplier:  G-Biosciences
An ELISA kit for the detection of TAFI (Mouse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.469ng/ml. 1 * 1 KIT
Catalog # GENOIT6007
Supplier:  G-Biosciences