ContactUsLinkComponent
An ELISA kit for the detection of GABA (Human). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 18.75pg/ml . 1 * 1 KIT
Catalog # GENOIT3980
Supplier:  G-Biosciences
An ELISA kit for the detection of GABA (Human). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 18.75pg/ml . 1 * 1 KIT
Catalog # GENOIT3980
Supplier:  G-Biosciences