Hematoxylin modified solution acc. Mayer
Catalog # GENO786-1265
Supplier: G-Biosciences
Hematoxylin modified solution acc. Mayer
Catalog # GENO786-1265
Supplier: G-Biosciences
CAS Number: 517-28-2
Jump to:
Product Details & Documents
Core component of Hematoxylin and Eosin (H&E) staining in pathology, research, and diagnostic laboratories.
- Manufactured with BSC certified reagents
Mayer’s Hemotoxylin Stain, modified for enhanced shelf-life while avoiding use of Chloral Hydrate.
Synonyms: DcH-48, Natural Black I, Hydroxybrasilin
- CAS Number:517-28-2
- Formula:C₁₆H₁₄O₆
- MDL Number:MFCD00078111
- Molecular Weight:302.28 g/mol



