ContactUsLinkComponent
ML (Modified Lowry) Protein Assay is compatible with a wide variety of detergents in addition to reagents such as EDTA and Tris. Sup plied with Non Animal Protein standard. 1,000 assays 1 * 1 KIT
Catalog # GENO786-1076
Supplier:  G-Biosciences
ML (Modified Lowry) Protein Assay is compatible with a wide variety of detergents in addition to reagents such as EDTA and Tris. Sup plied with Non Animal Protein standard. 1,000 assays 1 * 1 KIT
Catalog # GENO786-1076
Supplier:  G-Biosciences