ContactUsLinkComponent
An ELISA kit for the detection of MIF (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.469ng/ml. 1 * 1 KIT
Catalog # GENOIT1483
Supplier:  G-Biosciences
An ELISA kit for the detection of MIF (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.469ng/ml. 1 * 1 KIT
Catalog # GENOIT1483
Supplier:  G-Biosciences