ContactUsLinkComponent
An ELISA kit for the detection of TFPI (Horse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
Catalog # GENOIT1459
Supplier:  G-Biosciences
An ELISA kit for the detection of TFPI (Horse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
Catalog # GENOIT1459
Supplier:  G-Biosciences