ContactUsLinkComponent
An ELISA kit for the detection of IL1a (Horse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
Catalog # GENOIT1449
Supplier:  G-Biosciences
An ELISA kit for the detection of IL1a (Horse). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
Catalog # GENOIT1449
Supplier:  G-Biosciences