Order Entry
Export
Orders LinkContactUsLinkComponent
Nuclear Fast Red solution (Kernechtrot)

Nuclear Fast Red solution (Kernechtrot)

Catalog # GENO786-1053
Supplier:   G-Biosciences
Nuclear Fast Red solution (Kernechtrot)
Catalog # GENO786-1053
Supplier:  G-Biosciences
CAS Number:  6409-77-4
Nuclear Fast Red solution (Kernechtrot)
CAS Number:  6409-77-4
Price
48,10 €
Log in for availability
1 * 100 mL
Product Details & Documents

Conventional stain used for numerous histology and cytology procedures.

  • Enhanced stability formula ensures long shelf-life and consistent results

Ready to use formulation of 0.1% nuclear fast red in 5% aluminum sulfate, optimized for histology and cytology procedures where reproducible results are important.

Synonyms:  CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
  • CAS Number:6409-77-4
  • Formula:C₁₄H₈NNaO₇S
  • MDL Number:MFCD00078493
  • Molecular Weight:357.28 g/mol

Documents