Order Entry
Export
Orders LinkContactUsLinkComponent
Giemsa's Stain stock solution

Giemsa's Stain stock solution

  GENO786-1067
Giemsa's Stain stock solution
  GENO786-1067
 :  51811-82-6
Giemsa's Stain stock solution
 :  51811-82-6
 
148,00 €
Log in for availability
1 * 1 L
 

Conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology procedures. Ready to use formulation optimized for histology and cytology procedures where reproducible results are important.

  • This stock solution has been formulated for consistent staining and extended shelf-life

Classic differential stain with a broad spectrum of applications that include cell viability, staining metaphases to look for chromosomal aberrations, blood smears, as well as other histology/cytology applications. Use Giemsa’s azure eosin methylene blue solution for any of its classical applications with minimal troubleshooting due to its standardized recipe.

Synonyms:  Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
  • CAS Number:51811-82-6
  • MDL Number:MFCD00081642