Giemsa's Stain stock solution
Catalog # GENO786-1066
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology procedures. Ready to use formulation optimized for histology and cytology procedures where reproducible results are important.
- This stock solution has been formulated for consistent staining and extended shelf-life
Classic differential stain with a broad spectrum of applications that include cell viability, staining metaphases to look for chromosomal aberrations, blood smears, as well as other histology/cytology applications. Use Giemsa’s azure eosin methylene blue solution for any of its classical applications with minimal troubleshooting due to its standardized recipe.
Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
- CAS Number:51811-82-6
- MDL Number:MFCD00081642



