ContactUsLinkComponent
An ELISA kit for the detection of IAA (Human). This uses Sandwich ELISA, Double Antigen and has a sensitivity of 1.875ng/ml. 1 * 1 KIT
Catalog # GENOIT2651
Supplier:  G-Biosciences
An ELISA kit for the detection of IAA (Human). This uses Sandwich ELISA, Double Antigen and has a sensitivity of 1.875ng/ml. 1 * 1 KIT
Catalog # GENOIT2651
Supplier:  G-Biosciences