ContactUsLinkComponent
An ELISA kit for the detection of ET-1 (Guinea pig). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1403
Supplier:  G-Biosciences
An ELISA kit for the detection of ET-1 (Guinea pig). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1403
Supplier:  G-Biosciences