ContactUsLinkComponent
An ELISA kit for the detection of Bax (Guinea pig). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
  GENOIT1393
An ELISA kit for the detection of Bax (Guinea pig). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094ng/ml. 1 * 1 KIT
  GENOIT1393