ContactUsLinkComponent
An ELISA kit for the detection of GBD1 (Goat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml. 1 * 1 KIT
Catalog # GENOIT1379
Supplier:  G-Biosciences
An ELISA kit for the detection of GBD1 (Goat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.875pg/ml. 1 * 1 KIT
Catalog # GENOIT1379
Supplier:  G-Biosciences