ContactUsLinkComponent
An ELISA kit for the detection of HTRA1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Catalog # GENOIT3202
Supplier:  G-Biosciences
An ELISA kit for the detection of HTRA1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Catalog # GENOIT3202
Supplier:  G-Biosciences