ContactUsLinkComponent
An ELISA kit for the detection of HSPD1/HSP-60 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.938ng/m l. 1 * 1 KIT
Catalog # GENOIT2294
Supplier:  G-Biosciences
An ELISA kit for the detection of HSPD1/HSP-60 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.938ng/m l. 1 * 1 KIT
Catalog # GENOIT2294
Supplier:  G-Biosciences