ContactUsLinkComponent
An ELISA kit for the detection of IL-31 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1655
Supplier:  G-Biosciences
An ELISA kit for the detection of IL-31 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1655
Supplier:  G-Biosciences