Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
An ELISA kit for the detection of VSNL1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
GENOIT2818
:
G-Biosciences
An ELISA kit for the detection of VSNL1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
GENOIT2818
:
G-Biosciences