ContactUsLinkComponent
An ELISA kit for the detection of MNX1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.9pg/ml. 1 * 1 KIT
Catalog # GENOIT2809
Supplier:  G-Biosciences
An ELISA kit for the detection of MNX1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 46.9pg/ml. 1 * 1 KIT
Catalog # GENOIT2809
Supplier:  G-Biosciences