Order Entry
Export
Orders LinkContactUsLinkComponent
Thermal transfer printer labels, type B-423

Thermal transfer printer labels, type B-423

Printer labels, B-423, THT-51-423-10, Polyester, Permanent acrylic, Thermal transfer, White, i5100, 25×6 mm, 10000 labels/roll
Catalog # BRDY030235
Supplier:   Brady
Thermal transfer printer labels, type B-423
Catalog # BRDY030235
Supplier:  Brady
Thermal transfer printer labels, type B-423
Thermal transfer printer labels, type B-423

Printer labels, B-423, THT-51-423-10, Polyester, Permanent acrylic, Thermal transfer, White, i5100, 25×6 mm, 10000 labels/roll

Price
132,00 €
Log in for availability
1 * 1 Roll
Product Details & Documents

Polyester labels for thermal transfer printers.

  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon separate, not in the same enclosure as the tape/sleeves

This material is UL, CSA, and AGA approved with the respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.

Specifications
  • Printer model:
    i5100
  • Printing method:
    Thermal transfer
  • Type:
    B-423
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    25×6 mm
  • Material:
    Polyester
  • Seq:
    04
  • Model:
    THT-51-423-10
  • Packed:
    10000 labels/roll
  • Adhesive:
    Permanent acrylic