Warning signs
Warning of explosive atmosphere, Polyester, 87×100 mm
Catalog # BRDY810888
Supplier: Brady
Jump to:
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifications
- Description:Warning of explosive atmosphere
- Dimensions:87×100 mm
- Material:Polyester
- Seq:14