ContactUsLinkComponent
Stirrer guide with cone NS 29/32, made of presicion calibrated glass tube, for shaft Ø 16 mm, surface ground and polished, made of DURAN® tubing 1 * 1 items
Catalog # LENZ06301629
Stirrer guide with cone NS 29/32, made of presicion calibrated glass tube, for shaft Ø 16 mm, surface ground and polished, made of DURAN® tubing 1 * 1 items
Catalog # LENZ06301629