Orders LinkContactUsLinkComponent
Microplates, 384-well
Microplates, 384-well
LoBase, clear
Catalog # GREI788101_80
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 384-well
Catalog # GREI788101_80
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 384-well
Microplates, 384-well
LoBase, clear
Price
1 150,00 €
Log in for availability
1 * 80 items
Product Details & Documents
PS. With the exception of the Small Volume™, all 384-well PS microplates have rounded square wells, which combine the advantages of flexible working volume (10 to 130 µl) with reduced wicking and bubbling. Small Volume™ plates have round wells with a conical geometry, which allows a working volume of just 4 to 25 µl. All plates are supplied without lid, except for some sterile formats.
  • Flat bottom well profile
  • Choice of solid or µClear® bottom
  • Medium or high binding versions available
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifications
  • Description:
    LoBase, clear
  • Seq:
    24
  • Cat. No.:
    GREI788101_80
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact