ContactUsLinkComponent
Kit (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of mouse Kit in cell culture supe rnates, cell lysates, serum and plasma (heparin or EDTA). 1 * 1 KIT
  ABNOKA5210
 :  Abnova
Kit (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of mouse Kit in cell culture supe rnates, cell lysates, serum and plasma (heparin or EDTA). 1 * 1 KIT
  ABNOKA5210
 :  Abnova