ContactUsLinkComponent
Ldlr (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse LDLR in cell culture supe rnates, cell lysates, serum, plasma (heparin) and urine. 1 * 1 KIT
Catalog # ABNOKA5179
Supplier:  Abnova
Ldlr (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse LDLR in cell culture supe rnates, cell lysates, serum, plasma (heparin) and urine. 1 * 1 KIT
Catalog # ABNOKA5179
Supplier:  Abnova