Order Entry
Export
Orders LinkContactUsLinkComponent
DNA transfection reagent, HeLaFect™

DNA transfection reagent, HeLaFect™

HeLaFect™ transfection reagent, 250 - 500 transfections with 1 µg of DNA, 1.000 µl
Catalog # OZBIHF21000
Supplier:   OZ BIOSCIENCES
DNA transfection reagent, HeLaFect™
Catalog # OZBIHF21000
Supplier:  OZ BIOSCIENCES
CAS Number:  
DNA transfection reagent, HeLaFect™
DNA transfection reagent, HeLaFect™

HeLaFect™ transfection reagent, 250 - 500 transfections with 1 µg of DNA, 1.000 µl

CAS Number:  
Price
305,61 €
Log in for availability
1 * 1.000 µl
Product Details & Documents

HeLaFect™ transfection reagent is specifically developed for HeLa cell lineage transfection with high efficiency. It is a lipid-based reagent based on Tee-Technology (Triggered Endosomal Escape). The cationic design of the reagent allows high nucleic acid compaction for efficient transport into HeLa cells. This reagent is composed of lipids, leading to high viability, and is ready to use.

  • Highly efficient, more than 80% of transfected HeLa cells
  • Ready-to-use, no need for additional buffer
  • Low nucleic acid amount minimised toxicity
  • High level of nucleic acid compaction
  • Easy and straight forward protocol
  • Compatible with any culture medium, medium change not required

Store at –20°C.

Delivery information: Shipped at room temperature.

Specifications

Specifications

  • Pk:
    1.000 µl
  • Description:
    HeLaFect™ transfection reagent, 250 - 500 transfections with 1 µg of DNA
  • Cat. No.:
    OZBIHF21000
  • Cell specific:
    HeLa

Specifications