Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Agilent J+W DB-624 Intuvo GC column module, 25 m, 0.20 mm, 1.12 µm 1 * 1 items
Catalog # HEWL128-1324-INT
Supplier:
Agilent
Expand image
Agilent J+W DB-624 Intuvo GC column module, 25 m, 0.20 mm, 1.12 µm 1 * 1 items
Catalog # HEWL128-1324-INT
Supplier:
Agilent