VWR® pHenomenal® IDP pH Electrodes
pHenomenal® IDP 721-LS pH electrode, pHenomenal® IDP 721-LS, 0,000 - 14,000 (±0,004), -5…+100 °C, Glass, KCl 3 mol/L, refillable, Platinum wire, 12×120 mm, 1,5 m cable, IDP-plug
662-2298
These IDP (Intelligent Digital Probe) pH/temperature electrodes can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
- Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
- Proven reliability and accuracy
- Correct measurement results due to direct signal processing in the sensor and digital transmission of values
- All parameters are easily measured with cable lengths of up to 100 metres
- Same universal connector for all electrodes and probes
Temperature probe: NTC 30 kOhm
The electrodes can provide the following data shown on the meter's screen:
- Model name
- Serial number
- Calibration status (+++, ++, +, –, blank)
- Calibration history includes the last 10 calibrations, including buffers used, mV values, etc.
Delivery information: Includes digital pH electrode only.
Documents
- Connection:1,5 m cable, IDP-plug
- Ø×L:12×120 mm
- Type:pHenomenal® IDP 721-LS
- Description:pHenomenal® IDP 721-LS pH electrode
- Material:Glass
- Diaphr.:Platinum wire
- Electrolyte:KCl 3 mol/L, refillable
- Temp.:-5…+100 °C
- pH:0,000 - 14,000 (±0,004)



