Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol ≥96%, powder USP

Poly pail, 2,5 kg
Catalog # SPCMMA165-2.5KG
Supplier:   Spectrum Chemical
D-(-)-Mannitol ≥96%, powder USP
Catalog # SPCMMA165-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  69-65-8
D-(-)-Mannitol ≥96%, powder USP
D-(-)-Mannitol ≥96%, powder USP

Poly pail, 2,5 kg

CAS Number:  69-65-8
Price
868,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Mannitol, Powder, USP.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1,49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specification Test Results

  • Assay:
    96% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    2,5 kg

Specifications