0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
MODIFICATION KIT INNER DOOR 115 APT2 1 * 1 items
BIND8500-0131
:
Binder
MODIFICATION KIT INNER DOOR 115 APT2 1 * 1 items
BIND8500-0131
:
Binder