Orders LinkContactUsLinkComponent
Wipes, Amplitude™ Sigma™ Plus
Wipes, Amplitude™ Sigma™ Plus
Amplitude™ Sigma™ Plus wipes, flat stacked, White, 305×305 mm
Catalog # CONTC1E-1212
Supplier:   CONTEC
Wipes, Amplitude™ Sigma™ Plus
Catalog # CONTC1E-1212
Supplier:  CONTEC
Wipes, Amplitude™ Sigma™ Plus
Wipes, Amplitude™ Sigma™ Plus
Amplitude™ Sigma™ Plus wipes, flat stacked, White, 305×305 mm
Price
1 320,00 €
Log in for availability
1 * 2.700 items
Product Details & Documents
The Amplitude™ Sigma™ Plus wipes are strong and durable, compatible with most solvents, and have outstanding sorbency.
  • Hydroentangled cellulose/poylester provides excellent strength and durability
  • Low levels of particles and extractables
  • Variety of blended fibers enable excellent absorbing capacity
Amplitude™ Sigma™ Plus Wipes are constructed using a nonwoven fabric that is a hydroentangled blend of polyester and cellulose fibers. Wet or dry, the wipe maintains a high level of strength and durability.
Specially treated to reduce particles and fibers, it is recommended when low particle generation combined with sorbency is of high priority.
Specifications
  • Colour:
    White
  • Description:
    Amplitude™ Sigma™ Plus wipes, flat stacked
  • L×W:
    305×305 mm
  • Seq:
    02