Orders LinkContactUsLinkComponent
Speedglas™ Outer Protection Plates
Speedglas™ Outer Protection Plates
3M™ Speedglas™ Outer protection plate, 9100, scratch resistant, 527001
Catalog # MMMAH527001N
Supplier:   3M
Speedglas™ Outer Protection Plates
Catalog # MMMAH527001N
Supplier:  3M
Speedglas™ Outer Protection Plates
Speedglas™ Outer Protection Plates
3M™ Speedglas™ Outer protection plate, 9100, scratch resistant, 527001
Price
79,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Speedglas™ Outside protection plates are designed for use with 3M™ Speedglas™ welding helmets and filters to protect the welding filter from sparks and spatter.
  • Outer protection plate
  • Protect welding filters from welding sparks and spatter
The Speedglas™ Outside protection plates are a replacement part designed to be used when the original has been damaged or worn out. Help protect welding filters from sparks and spatter by ensuring that the outside protection plate is functioning properly.
Specifications
  • Description:
    3M™ Speedglas™ Outer protection plate, 9100, scratch resistant, 527001
  • Seq:
    02