Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Anti-CD40L antibody [24-31] (Phycoerythrin) 1 * 100 Tests
Catalog # ABCAAB234220-100
Supplier:
Abcam
Expand image
Anti-CD40L antibody [24-31] (Phycoerythrin) 1 * 100 Tests
Catalog # ABCAAB234220-100
Supplier:
Abcam