Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE BOTTLE WEITHALS 1000ML 1 * 1 items
Catalog # NEUB0863-15-1045
Supplier:
NEUBERT VOLUME GLASSWAERE
Expand image
STORAGE BOTTLE WEITHALS 1000ML 1 * 1 items
Catalog # NEUB0863-15-1045
Supplier:
NEUBERT VOLUME GLASSWAERE