Capsule counting trays, SteriWare®
Sterile, White, 260×130×10 mm
Supplier: Sampling Systems
Jump to:
Product Details & Documents
The disposable counting trays are made from robust, shatterproof PS. They are suitable for use with all capsules from size 2 to 00.
- BSE/TSE free
- Suitable for capsules size 2 to size 00
- Manufactured and packed in a cleanroom
- Can be used straight from the bag
Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Packaging: Individually bagged in a heat-sealed laminate bag.
Documents
Specifications
- Colour:White
- Description:Sterile
- Seq:02
- W×D×H:260×130×10 mm