Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
H+H FlexShelf-Fachb. Kunststoff 400x600 Kunststoff grau, 19 mm BxTxH=443x627x19 mm 1 * 1 ST
Catalog # HHSY7532455-46-19
Supplier:
H H SYSTEMS
Expand image
H+H FlexShelf-Fachb. Kunststoff 400x600 Kunststoff grau, 19 mm BxTxH=443x627x19 mm 1 * 1 ST
Catalog # HHSY7532455-46-19
Supplier:
H H SYSTEMS