Safety ankle boots, lace-up, Basic Mid, Jori
37, Black
Catalog # ELTN16331/37
Supplier: ELTEN
Safety ankle boots, lace-up, Basic Mid, Jori
Catalog # ELTN16331/37
Supplier: ELTEN
Jump to:
Product Details & Documents
Extremely lightweight safety ankle boot made from cowhide leather, reflective materials, skin friendly textile lining, a full padded bellows tongue plus collar padding and a dual-density PU/PU jo_Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
- Climate-regulating, breathable textile lining
- Full length inlay sole
- PU scuff cap protects the upper material
- Puncture resistant steel midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
: EN ISO 20345; S3, SRC
Specifications
- Shoe size:37
- Colour:Black
- Cat. No.:ELTN16331/37
- Environmental certification:ISO 14001
- Environmentally Preferable:Low Manufacturing Impact