Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dibromoadamantane

Bottle, 500 g
Catalog # APOSOR315789-500G
Supplier:   Apollo Scientific
1,3-Dibromoadamantane
Catalog # APOSOR315789-500G
Supplier:  Apollo Scientific
CAS Number:  876-53-9
1,3-Dibromoadamantane

Bottle, 500 g

CAS Number:  876-53-9
Price
356,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:876-53-9
  • Formula:C₁₀H₁₄Br₂
  • MDL Number:MFCD00077218
  • Molecular Weight:294.03 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications