0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Dibromoadamantane
1,3-Dibromoadamantane
1,3-Dibromoadamantane
1,3-Dibromoadamantane
Bottle, 25 g
APOSOR315789-25G
:
Apollo Scientific
1,3-Dibromoadamantane
APOSOR315789-25G
:
Apollo Scientific
:
876-53-9
Expand image
1,3-Dibromoadamantane
Bottle, 25 g
Add to cart
:
876-53-9
31,90 €
Unit of Measure
1 * 25 g
-
+
Add to cart
:
876-53-9
:
C₁₀H₁₄Br₂
:
MFCD00077218
:
294.03 g/mol
:
Ambient
Pack type:
Bottle
Pk:
25 g